Inserisci ordine
Italy
ContactUsLinkComponent
6 components 125mg/l each of Au Hg Mo Sb Sn Ti in HCl 8% . Duree de conservation : 12 mois a ouverture du flacon, Duree de validite: 24 mois. Store under normal laboratory conditions, at temperatures between 15 to 25C 1 * 100 mL
Catalog # CMPLRM053268L1
6 components 125mg/l each of Au Hg Mo Sb Sn Ti in HCl 8% . Duree de conservation : 12 mois a ouverture du flacon, Duree de validite: 24 mois. Store under normal laboratory conditions, at temperatures between 15 to 25C 1 * 100 mL
Catalog # CMPLRM053268L1
Frequently Bought Together