Order Entry
Italy
ContactUsLinkComponent
CHROMABOND Multi 96 HR-XCW monoblock - weak mixed-mode polymer-based cation exchanger - particle size: 45 µm contents of sorbent: 2 5 mg material: PP consists of: 96 well plate 1 * 1 items
Catalog # MANA738570.025M
Supplier:  MACHEREY-NAGEL
CHROMABOND Multi 96 HR-XCW monoblock - weak mixed-mode polymer-based cation exchanger - particle size: 45 µm contents of sorbent: 2 5 mg material: PP consists of: 96 well plate 1 * 1 items
Catalog # MANA738570.025M
Supplier:  MACHEREY-NAGEL
Frequently Bought Together