Order Entry
Italy
ContactUsLinkComponent
CHROMABOND QuEChERS Mix XI C18 ec clean-up mix consisting of: - 150 mg MgSO4 - 50 mg CHROMABOND C18 ec individually weighed and fil led into 2 mL centrifuge tube with snap cap pack of 50 1 * 50 items
Catalog # MANA730983.2
Supplier:  MACHEREY-NAGEL
CHROMABOND QuEChERS Mix XI C18 ec clean-up mix consisting of: - 150 mg MgSO4 - 50 mg CHROMABOND C18 ec individually weighed and fil led into 2 mL centrifuge tube with snap cap pack of 50 1 * 50 items
Catalog # MANA730983.2
Supplier:  MACHEREY-NAGEL
Frequently Bought Together