Combipack consisting of 702885: 1.5 mL Crimp Neck Vial N 11 outer diameter: 11.6 mm, outer height: 32 mm clear, flat bottom, wide o pening, label + scale and 70256: N 11 Aluminium crimp cap, silver, center hole Natural Rubber/Butyl red-orange/TEF col 1 * 1.000 items
Catalog # MANA702087
Supplier: MACHEREY-NAGEL
Combipack consisting of 702885: 1.5 mL Crimp Neck Vial N 11 outer diameter: 11.6 mm, outer height: 32 mm clear, flat bottom, wide o pening, label + scale and 70256: N 11 Aluminium crimp cap, silver, center hole Natural Rubber/Butyl red-orange/TEF col 1 * 1.000 items
Catalog # MANA702087
Supplier: MACHEREY-NAGEL
Frequently Bought Together
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Test Sieves, 200×50 mm
Supplier: VWR Collection
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
SSL Broth and OFS Agar
Supplier: DOEHLER GROUP
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
VWR®, Crimp Caps for Crimp Neck Vials ND11
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection