Test tube racks, holes 3- row racks, 36 holes, diam 18 mm each (transverses for sizes 15 and 24) 1 * 1 items
MEMMB48834WTB6
: MEMMERT
Test tube racks, holes 3- row racks, 36 holes, diam 18 mm each (transverses for sizes 15 and 24) 1 * 1 items
MEMMB48834WTB6
: MEMMERT
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection