MobileALERT, notification by SMS in case of any error or alarm of the device (requires option H6) 1 * 1 items
MEMMICO105C3
: MEMMERT
MobileALERT, notification by SMS in case of any error or alarm of the device (requires option H6) 1 * 1 items
MEMMICO105C3
: MEMMERT
Frequently Bought Together
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection