Screw caps for SuperClear® microtubes
White
Catalog # 3600-871-000-9
Supplier: Labcon
Jump to:
Product Details & Documents
Caps only.
- Endotoxin Free (non-pyrogenic)
- Polyethylene USP Class VI resin
- Polypropylene
- Autoclavable
Specifications
- Colour:White
- Seq:08
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



