Order Entry
Italy
Orders LinkContactUsLinkComponent
Accessories for ASPEC® 241 System

Accessories for ASPEC® 241 System

Tubes glass, 12×75 mm, 6 ml, 250/pack
Catalog # GILI2954726
Supplier:   GILSON LCMS
Accessories for ASPEC® 241 System
Catalog # GILI2954726
Supplier:  GILSON LCMS
Accessories for ASPEC® 241 System
Accessories for ASPEC® 241 System

Tubes glass, 12×75 mm, 6 ml, 250/pack

Price
102,00 €
Log in for availability
1 * 250 Vial
Product Details & Documents
Specifications
  • Description:
    Tubes glass, 12×75 mm, 6 ml, 250/pack
  • Seq:
    26
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.