Accessories for ASPEC® 241 System
Rack, code 330, holds 60 12×32 mm vials (2 ml)
Catalog # GILI260440031
Supplier: GILSON LCMS
Jump to:
Product Details & Documents
Specifications
- Description:Rack, code 330, holds 60 12×32 mm vials (2 ml)
- Seq:13



