Order Entry
Italy
Orders LinkContactUsLinkComponent
Phosphorus ICP standards, (CRMs), Fluka™

Phosphorus ICP standards, (CRMs), Fluka™

Phosphorus, P, 1000 mg/L, 0,05% H₂SO₄, Plastic bottle, 100 mL
Catalog # HONCE1114-100ML
Supplier:   Honeywell Chemicals
Phosphorus ICP standards, (CRMs), Fluka™
Catalog # HONCE1114-100ML
CAS Number:  
Phosphorus ICP standards, (CRMs), Fluka™

Phosphorus, P, 1000 mg/L, 0,05% H₂SO₄, Plastic bottle, 100 mL

CAS Number:  
Price
77,57 €
Log in for availability
1 * 100 mL
Product Details & Documents

Certified Reference Materials (CRMs)

  • Traceable to SRM from NIST

 : ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).

Specifications

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    0,05% H₂SO₄
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Phosphorus
  • Symbol:
    P

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.