Order Entry
Italy
Orders LinkContactUsLinkComponent
Affinity chromatography columns, StrepTrap™ XT

Affinity chromatography columns, StrepTrap™ XT

StrepTrap™ XT, PP, 34 µm, 7,00 mm, 25 mm, 5×1 ml, 1 ml/min
Catalog # 29-4013-20
Supplier:   Cytiva
Affinity chromatography columns, StrepTrap™ XT
Catalog # 29-4013-20
Supplier:  Cytiva
Affinity chromatography columns, StrepTrap™ XT
Affinity chromatography columns, StrepTrap™ XT

StrepTrap™ XT, PP, 34 µm, 7,00 mm, 25 mm, 5×1 ml, 1 ml/min

Price
615,00 €
Log in for availability
1 * 5 pezzi
Product Details & Documents

A chromatography resin for the purification of recombinant proteins tagged with Strep-tag® II and Twin-Strep-tag®, which provides higher affinity, resulting in higher yields and lower loss of valuable target protein.

  • Highly pure Strep-tag® II and Twin-Strep-tag® recombinant proteins
  • Fast purifications with simple elution
  • Physiological conditions and mild elution preserve target protein activity
  • Fast and easy regeneration

Compatible with a wide range of detergents, denaturants, and other additives. Available in 10 and 50 ml lab packs of bulk resin and prepacked in 1 and 5 ml StrepTrap™ XT chromatography columns for convenience.

Specifications
  • Flow rate:
    1 ml/min
  • Ø int.:
    7,00 mm
  • Particle Size:
    34 µm
  • Capacity:
    5×1 ml
  • Description:
    StrepTrap™ XT
  • Length:
    25 mm
  • Column hardware:
    PP
  • Cat. No.:
    29-4013-20
  • Max. pressure:
    5 bar
  • Storage:
    2...8 °C in 20% ethanol
  • pH range:
    6 - 10
  • Application:
    Purification of recombinant proteins tagged with Strep-tag® II and Twin-Strep-tag®
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.