Please speak with Hichrom ChromSupport each time before quoting or ordering this item. This is a custom packed column and is non-returnable. Part Number: C-5000-11 Stationary Phase: 5% CBWX 20M on Chromosorb T 40/60, Preconditioned: Yes Hardware: Sta 1 * 1 items
Catalog # HISMC-5000-11
Supplier: HICHROM
Frequently Bought Together