Sliding Drawer Racks for Upright Freezer holds 24x 2ml plastic boxes (4x6) 1 * 1 items
Catalog # SANYSDR-624-P
Supplier: PHC Corporation
Sliding Drawer Racks for Upright Freezer holds 24x 2ml plastic boxes (4x6) 1 * 1 items
Catalog # SANYSDR-624-P
Supplier: PHC Corporation
Frequently Bought Together
VWR® Pipette Stands
Supplier: VWR Collection
VWR® Ergonomic High Performance (EHP), Single Channel Pipettes, Mechanical, Variable Volume
Supplier: VWR Collection
VWR®, Digital Timer, One Channel
Supplier: VWR Collection
VWR® Traceable® Digital Thermo-Hygrometer
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
Accessories for marco
Supplier: Brand
VWR®, Digital Timer, Three Channels, with Clock
Supplier: VWR Collection
VWR®, Digital Timer, One Channel
Supplier: VWR Collection
Accessories for water purification systems, arium®
Supplier: Sartorius
VWR® Pipette Filler, White/Blue
Supplier: VWR Collection
VWR® Pipette Controller
Supplier: VWR Collection
VWR®, Digital Timer, One Channel
Supplier: VWR Collection
Digital timer
Supplier: GLASWARENFABRIK KARL HECHT
Pipette stand, linear
Supplier: Sartorius
Eppendorf® Pipette Holder System
Supplier: EPPENDORF
VWR®, Digital Timer, One Channel
Supplier: VWR Collection
VWR®, Digital Timer, Four Channels, with Clock
Supplier: VWR Collection
Cartridges for Milli-Q®
Supplier: Merck
DNA Ligase T4 5x1000 units (5u/µl) 1 * 1 KIT
Supplier: Thermo Fisher Scientific