Order Entry
Italy
Orders LinkContactUsLinkComponent
 

SteriWare® Liquid Funnels

Funnel, natural, sterile, 8 mm, 50 mm
  300-0591
SteriWare® Liquid Funnels
  300-0591
SteriWare® Liquid Funnels
SteriWare® Liquid Funnels

Funnel, natural, sterile, 8 mm, 50 mm

 
283,00 €
   
1 * 100 pezzi
 

Single use, disposable liquid funnels ideal for laboratories and manufacturing areas. Designed to use straight from bags.

  • Made of HDPE
  • Moulded and packed in a class 100,000 cleanroom
  • With small hygenic side handle
  • BSE/TSE free
  • Available sterile (gamma irradiated)

 : Material conforms to FDA, EU 10/2011 and EC 1935/2004. Full batch traceability.

 : Individually bagged.

 
  • Description:
    Funnel, natural, sterile
  • Seq:
    02
  • Int.Ø top:
    50 mm
  • Stem O-Ø:
    8 mm