Disulfoton-oxon-sulfone 1 * 10 mg
Catalog # EHERCA12982200
Supplier: EHRENSTORFER
Frequently Bought Together
Conductivity standards, Certipur® Supelco®
Supplier: Merck
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Titanium(III) chloride 15% in hydrochloric acid 10% Reag. Ph. Eur. 1091201
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Buffer solutions in sachets
Supplier: Mettler - Toledo
VWR®, Primary colour solutions, Reagent Ph. Eur.
Supplier: VWR Chemicals
VWR®, General purpose viscosity standards
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
Conductivity standards
Supplier: Mettler - Toledo
Buffer solutions, according to EP and USP, Certipur®
Supplier: Merck
pH buffers, ready to use
Supplier: WTW
Buffer solutions
Supplier: REAGECON
Lead acetate cotton Reag. Ph. Eur. 1048101
Supplier: VWR Chemicals