N-(carboxymethyl)-N,N-dimethyl-3-((((perfluorohexyl)ethyl)sulfonyl)amino)-, hydroxide, inner salt 1 * 50 mg
Catalog # EHERC11041300
Supplier: EHRENSTORFER
N-(carboxymethyl)-N,N-dimethyl-3-((((perfluorohexyl)ethyl)sulfonyl)amino)-, hydroxide, inner salt 1 * 50 mg
Catalog # EHERC11041300
Supplier: EHRENSTORFER
Frequently Bought Together
Supelco® TraceCERT® Organic Reference Standard, Tebutam
Supplier: Merck
Supelco® Organic Reference Standard, Oxalic Acid
Supplier: Merck
Cyclaniliprole 1 * 10 mg
Supplier: EHRENSTORFER
VWR®, Cone and Plate viscosity standards
Supplier: VWR Chemicals
Potassium dihydrogen phosphate 0.2 mol/l in aqueous solution Reag. Ph. Eur. 1069601
Supplier: VWR Chemicals
Mesotrione Analytical standard, Fluka™
Supplier: Honeywell Chemicals
Sulphanilic acid 0.02 mol/l in acetic acid (25%) Reag. Ph. Eur. 1086203
Supplier: VWR Chemicals
Conductivity standard 1413 µS/cm (25°C; 0,01 mol/L; KCl) for conductivity measurement
Supplier: SI Analytics
Complete Volatiles Kit - WP, Supelco®
Supplier: Merck
Linezolid, Reference Standards
Supplier: USP
LOFEXIDINE HCL 1 * 1 mL
Supplier: Merck
Praziquantel certified 1 * 0,25 g
Supplier: EHRENSTORFER
Supelco® PESTANAL® Organic Reference Standard, Molinate
Supplier: Merck
Butachlor certified 1 * 0,1 g
Supplier: EHRENSTORFER