PIANO Olefins Mixture 92 1 * 1 mL
Catalog # EHERGA0900092
Supplier: EHRENSTORFER
Frequently Bought Together
Sucrose solutions for refractometer calibration
Supplier: BELLINGHAM STANLEY
Reagents for alkalinity determination using Lovibond® MD photometers
Supplier: Lovibond
Phthalic anhydride
Supplier: EHRENSTORFER
Triafamone 1 * 50 mg
Supplier: EHRENSTORFER
Multi-element calibration standard 5
Supplier: PerkinElmer
Sudan orange G
Supplier: EHRENSTORFER
Trifluoroacetic acid 1 * 1 mL
Supplier: EHRENSTORFER
Glufosinate-N-acetyl 1 * 10 mg
Supplier: EHRENSTORFER
Alpha Tocopheryl Acid Succinate, Reference Standard
Supplier: USP
Eicosapentaenoic Acid, Reference Standards
Supplier: USP
Chlorthiophos certified 1 * 0,1 g
Supplier: EHRENSTORFER
Barium standard solution, 1000 mg/L Ba in 5% nitric acid (from Ba(NO₃)₂), standard for ICP, Specpure®
Supplier: Thermo Fisher Scientific
Supelco® PESTANAL® Organic Reference Standard, Hexazinone
Supplier: Merck
Ampicillin, Reference Standards
Supplier: USP
Roxithromycin certified 1 * 0,1 g
Supplier: EHRENSTORFER