Order Entry
Italy
Orders LinkContactUsLinkComponent
VWR®, Flash point reference standard, Cleveland Open Cup

VWR®, Flash point reference standard, Cleveland Open Cup

Flash Point Reference Standard, Cleveland Open Cup, 164 °C, ASTM D92, Glass bottle, 3×80 ml, 1 SET
Codice catalogo 99162.001
Nome del fornitore:   VWR Chemicals
VWR®, Flash point reference standard, Cleveland Open Cup
Codice catalogo 99162.001
Nome del fornitore:  VWR Chemicals
Numero CAS:  
VWR®, Flash point reference standard, Cleveland Open Cup
VWR®, Flash point reference standard, Cleveland Open Cup

Flash Point Reference Standard, Cleveland Open Cup, 164 °C, ASTM D92, Glass bottle, 3×80 ml, 1 SET

Numero CAS:  
Price
232,00 €
Log in for availability
1 * 1 SET
Product Details & Documents

COC flash point standards are tested and certified in accordance with ASTM D92 'Standard Test Method for Flash and Fire Points by Cleveland Open Cup Tester.'

  • Certified according to ISO 17025/ISO 17034 under UKAS accreditation
  • One bottle per test ensures zero contamination/high-end volatile loss
  • Fully traceable to international standards
  • Supplied with MSDS
  • Tamper-evident security packaging
  • 12 month shelf life

These UKAS certified standards are specifically designed to be used for regular instrument performance checks. Packed uniquely in 3×80 ml 'single shot' bottles ensures the highest level of sample integrity. An economical, time saving addition to any laboratory concerned with the generation of valid analytical test data. Typically used in, but not limited to, the analysis of petroleum products with flash points above 79 °C (175 °F) and below 400 °C (752 °F).

Certifications: UKAS ISO 17025 and ISO 17034.

Caution: The flash point given in the ordering table below is the nominal flash point - actual batch values are given in the certification documents supplied with the product and also on the label.

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 SET
  • Description:
    Flash Point Reference Standard, Cleveland Open Cup
  • Pk Info:
    3×80 ml
  • Flash point:
    164 °C
  • Test method:
    ASTM D92

Specifications