10 reactions-The Thermo Scientific SureQuant AKT Pathway (Phospho) IP and MS Sample Preparation Module provides a complete reagent s olution for the multiplexed enrichment and sample preparation for studies of key phosphorylation events of the AKT si 1 * 10 Assays
Catalog # PIERA40086
Supplier: Thermo Fisher Scientific
Frequently Bought Together