Neodym-Magnet 10x3 1 * 1 items
Catalog # SHPS5005-10000-172
Supplier: SHP Steriltechnik
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Schutzfilter 5 ml für Eppendorf Research Plus 1 * 50 ST
Supplier: EPPENDORF
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Accessories for pipetus® / pipetus®-standard / pipetus®-junior
Supplier: HIRSCHMANN
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Accessories for accu-jet® pro
Supplier: Brand
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Accessories for Transferpette® electronic pipettes
Supplier: Brand
VWR®, Accessories for Mechanical Pipettes
Supplier: VWR Collection
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Laboratory bottles, narrow neck, with screw cap, DURAN®
Supplier: DWK Life Sciences
Vacuum seal PTFE / FKM
Supplier: Heidolph Instruments GmbH & Co.KG
BIOTECTOR ACID REAGENT WITH CATALYST 1 * 20 L
Supplier: Hach