Order Entry
Italy
ContactUsLinkComponent
Thermometer similar to ASTM 110F, solid stem, 272,5+277,5:0,1F, white backed, capillary tube specially coated inside, with non-wett ing blue special liquid, 300-310x6-8mm, total immersion, top finish round, durable pigment, with ice point scale, wit 1 * 1 items
Catalog # AMARA301480-CB
Supplier:  Amarell
Thermometer similar to ASTM 110F, solid stem, 272,5+277,5:0,1F, white backed, capillary tube specially coated inside, with non-wett ing blue special liquid, 300-310x6-8mm, total immersion, top finish round, durable pigment, with ice point scale, wit 1 * 1 items
Catalog # AMARA301480-CB
Supplier:  Amarell
Frequently Bought Together