Order Entry
Italy
ContactUsLinkComponent
Led light cubes DAPI - EX:385/30 EM:450/50 DM:425 1 * 1 items
Catalog # ECHLCUBE4001
Supplier:  DISCOVER ECHO
Led light cubes DAPI - EX:385/30 EM:450/50 DM:425 1 * 1 items
Catalog # ECHLCUBE4001
Supplier:  DISCOVER ECHO