Order Entry
Italy
ContactUsLinkComponent
Troemner OIML Precision Weights Weight Set(8), 50 g-1 g, OIML E2, NVLAP 1 * 1 items
Catalog # OHAU80780384
Supplier:  OHAUS
Troemner OIML Precision Weights Weight Set(8), 50 g-1 g, OIML E2, NVLAP 1 * 1 items
Catalog # OHAU80780384
Supplier:  OHAUS
Frequently Bought Together