MIKROKUGELN BEREICH 072 MAGPLEX-C 1 * 1 mL
Catalog # MSPPMC10072-01
Supplier: MARKET SOURCE PART PROCESS
MIKROKUGELN BEREICH 072 MAGPLEX-C 1 * 1 mL
Catalog # MSPPMC10072-01
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together
Paraffin, Solidification point: 54 - 56 °C, Q Path® for microscopy
Supplier: VWR Chemicals
pH indicator papers, MQuant®, Supelco®
Supplier: Merck
Sodium hydroxide, pellets, pure, Supelco®
Supplier: Merck
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Test strips for peracetic acid, MQuant®, Supelco®
Supplier: Merck
Formaldehyde 4% stabilised, buffered (pH 7.0 ±0.1)
Supplier: VWR Chemicals
Ammonia solution 28%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
VWR® Rota®, pH indicator paper, 1 to 14
Supplier: VWR Chemicals
Visual colour kits, sulphite, MQuant®, Supelco®
Supplier: Merck
Test Strips, nitrate, MQuant®, Supelco®
Supplier: Merck
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
Supplier: Merck
Immersion oil for microscopy, Sigma-Aldrich®
Supplier: Merck
Test strips for ammonium, MQuant®, Supelco®
Supplier: Merck
Test strips for peroxide, MQuant®, Supelco®
Supplier: Merck
Test strips, peroxide, Dosatest
Supplier: VWR Chemicals
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals