Racks for Axygen® MultiRack tips
MultiRack empty rack, 5/pack, 1 pack/case, 200 µl extended length, 300 µl extended length, and 1000 µl extended length tips
Supplier: Corning
Jump to:
Product Details & Documents
Documents
Specifications
- Description:MultiRack empty rack, 5/pack, 1 pack/case
- For use with:200 µl extended length, 300 µl extended length, and 1000 µl extended length tips