Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
ACHIRAL ANALYTICAL COLUMN DAICEL DCpak® PTZ 150 x 4,6MM 5 micron 1 * 1 items
Catalog # DAICAC324
Supplier:
CHIRAL TECHNOLOGIES
Expand image
ACHIRAL ANALYTICAL COLUMN DAICEL DCpak® PTZ 150 x 4,6MM 5 micron 1 * 1 items
Catalog # DAICAC324
Supplier:
CHIRAL TECHNOLOGIES
Frequently Bought Together
Viral inactivation solution, Biotech Reagent, J.T.Baker®
Supplier: Avantor
Avantor® ACE® UltraCore SuperPhenylHexyl, HPLC/UHPLC Columns, 2,5 µm
Supplier: Avantor
Supelco® BIOshell™ IgG 1000 Å C4 Fused-Core® HPLC Columns
Supplier: Merck
Blank Vespel Ferrule Thermo MS Interface 1 * 10 items
Supplier: Thermo Fisher Scientific
VWR® Hichrom Nuts and Ferrules
Supplier: Avantor
Accessories for sample concentrators, Reacti Vap™
Supplier: Thermo Fisher Scientific