Order Entry
Italy
ContactUsLinkComponent
8 components As 2000mg/l Co 2000mg/l Cr 5000mg/l Cu 5000mg/l Ni 5000mg/l Pb 5000mg/l V 5000mg/l Zn 10000mg/l in HNO3 5% . Duree de conservation : 12 mois a ouverture du flacon, Duree de validite: 24 mois. Store under normal laboratory 1 * 100 mL
Catalog # CMPLRM004303L1
8 components As 2000mg/l Co 2000mg/l Cr 5000mg/l Cu 5000mg/l Ni 5000mg/l Pb 5000mg/l V 5000mg/l Zn 10000mg/l in HNO3 5% . Duree de conservation : 12 mois a ouverture du flacon, Duree de validite: 24 mois. Store under normal laboratory 1 * 100 mL
Catalog # CMPLRM004303L1