O RING AND SEAL GUIDE P20G/P20L 1 * 5 items
Catalog # GILSFA07013
Supplier: GILSON
Frequently Bought Together
BIOTECTOR BASE REAGENT 1 * 20 L
Supplier: Hach
Bottom wired shelf, stainless steel for IL 150R/IL 250R 1 * 1 items
Supplier: VWR Collection
VALVE HVXM 3-5 1 * 1 items
Supplier: HAMILTON BONADUZ
POWER SUPPLY 230 VAC 1 * 1 items
Supplier: THERMO H + P LABORTECHNIK
Threaded cap 1 * 1 items
Supplier: IKA
CAROUSEL KIT, METAL FOR 10 BOTTLES Ø35MM 1 * 1 items
Supplier: UVP ULTRA VIOLET PRODUCTS
STERICULT FILTER KIT 3307/3308/3310/3311 1 * 1 items
Supplier: Thermo Scientific
BIOTECTOR ACID REAGENT WITH CATALYST 1 * 20 L
Supplier: Hach
SAFETY VALVE 1 * 1 items
Supplier: CERTOCLAV
PCBA MTS 2/4 Shaker Motor Control 230V 1 * 1 items
Supplier: IKA
SHAFT KIT UHP 10 W/SEAL+ORING 1 * 1 KIT
Supplier: VWR Collection
O RING AND SEAL GUIDE P1000G/P1000L 1 * 5 items
Supplier: GILSON