Order Entry
Italy
ContactUsLinkComponent
CHLOROFORM-D, FOR NMR, CONT. 0.03V/V% TMS,IN 0.75 ML AMP.,99.8+ ATOM % D 1 * 7,5 mL
Catalog # ACRO464020075
CHLOROFORM-D, FOR NMR, CONT. 0.03V/V% TMS,IN 0.75 ML AMP.,99.8+ ATOM % D 1 * 7,5 mL
Catalog # ACRO464020075
Frequently Bought Together