Order Entry
Italy
ContactUsLinkComponent
Rabbit IgG polyclonal antibody for FGF2 detection. Tested with WB, Direct ELISA in Mouse Rat. 1 * 100 µG
Catalog # BSBTA00121-2
Supplier:  Boster Bio
Rabbit IgG polyclonal antibody for FGF2 detection. Tested with WB, Direct ELISA in Mouse Rat. 1 * 100 µG
Catalog # BSBTA00121-2
Supplier:  Boster Bio
Frequently Bought Together