A ELISA kit containing the core components for developing solid phase sandwich ELISA assay to quantitatively detects Oncostatin M/OS M in Mouse samples. The kit contains antibody pairs, recombinant protein standards and HRP conjugated ABC. 1 * 1 KIT
Catalog # BSBTEZ0479
Supplier: Boster Bio
Frequently Bought Together