Order Entry
Italy
ContactUsLinkComponent
Detect Human Antithrombin Iii/SERPINC1 with <10pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, serum, plasma (heparin, EDTA, citrate) and urine. 1 * 1 KIT
Catalog # BSBTFEK1393
Supplier:  Boster Bio
Detect Human Antithrombin Iii/SERPINC1 with <10pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, serum, plasma (heparin, EDTA, citrate) and urine. 1 * 1 KIT
Catalog # BSBTFEK1393
Supplier:  Boster Bio
Frequently Bought Together