Order Entry
Italy
ContactUsLinkComponent
Rabbit IgG polyclonal antibody for Loricrin/LOR detection. Tested with WB, IHC-P, Direct ELISA in Human Mouse Rat. 1 * 100 µG
Catalog # BSBTA05290-1
Supplier:  Boster Bio
Rabbit IgG polyclonal antibody for Loricrin/LOR detection. Tested with WB, IHC-P, Direct ELISA in Human Mouse Rat. 1 * 100 µG
Catalog # BSBTA05290-1
Supplier:  Boster Bio
Frequently Bought Together