Order Entry
Italy
ContactUsLinkComponent
Rabbit IgG polyclonal antibody for EIF4A1/2/3 detection. Tested with WB, IHC-P, ICC/IF, FCM, Direct ELISA in Human Mouse Rat. 1 * 100 µG
Catalog # BSBTA03922-2
Supplier:  Boster Bio
Rabbit IgG polyclonal antibody for EIF4A1/2/3 detection. Tested with WB, IHC-P, ICC/IF, FCM, Direct ELISA in Human Mouse Rat. 1 * 100 µG
Catalog # BSBTA03922-2
Supplier:  Boster Bio
Frequently Bought Together