Order Entry
Italy
ContactUsLinkComponent
Rabbit IgG Polyclonal Anti-Human ADO Antibody DyLight&reg 488 Conjugated, Flow Validated. 1 * 100 µG
Catalog # BSBTA02700-DYL488
Supplier:  Boster Bio
Rabbit IgG Polyclonal Anti-Human ADO Antibody DyLight&reg 488 Conjugated, Flow Validated. 1 * 100 µG
Catalog # BSBTA02700-DYL488
Supplier:  Boster Bio
Frequently Bought Together