Rabbit IgG polyclonal antibody for SMARCA2/BRM detection. Tested with WB, FCM, Direct ELISA in Human Mouse Rat. 1 * 100 µG
BSBTA01888-2
Rabbit IgG polyclonal antibody for SMARCA2/BRM detection. Tested with WB, FCM, Direct ELISA in Human Mouse Rat. 1 * 100 µG
BSBTA01888-2
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection