Order Entry
Italy
ContactUsLinkComponent
Rabbit IgG polyclonal antibody for Metabotropic Glutamate Receptor 5/GRM5 detection. Tested with WB, Direct ELISA in Human Mouse Rat . 1 * 100 µG
Catalog # BSBTA01338-1
Supplier:  Boster Bio
Rabbit IgG polyclonal antibody for Metabotropic Glutamate Receptor 5/GRM5 detection. Tested with WB, Direct ELISA in Human Mouse Rat . 1 * 100 µG
Catalog # BSBTA01338-1
Supplier:  Boster Bio
Frequently Bought Together