Order Entry
Italy
ContactUsLinkComponent
Volumetric pipette 12,5 ml, Class AS, blue graduated, Tolerance: ±0,020 ml, Color-Code: -special size volumetric pipettes, tolerances on the basis of DIN EN ISO 648, inclusive batch certificate 1 * 12 items
Catalog # WITG3.535.130.BL
Supplier:  witeg Labortechnik
Volumetric pipette 12,5 ml, Class AS, blue graduated, Tolerance: ±0,020 ml, Color-Code: -special size volumetric pipettes, tolerances on the basis of DIN EN ISO 648, inclusive batch certificate 1 * 12 items
Catalog # WITG3.535.130.BL
Supplier:  witeg Labortechnik
Frequently Bought Together