Order Entry
Italy
Orders LinkContactUsLinkComponent
Reagent reservoirs
Reagent reservoirs
Reagent reservoir, PVC, 55 ml, -
Catalog # HEAT120753
Supplier:   HEATHROW
Reagent reservoirs
Catalog # HEAT120753
Supplier:  HEATHROW
Reagent reservoirs
Reagent reservoirs
Reagent reservoir, PVC, 55 ml, -
Price
90,90 €
Log in for availability
1 * 100 pezzi
Product Details & Documents
These disposable reagent reservoirs have a fresh and modern design with enhanced features to streamline pipetting procedures.
  • Trough design with a 'v' shaped bottom helps ensure that all the reagent is accessible and can be easily aspirated
  • Thick wall construction and wide base adds stability and durability making the reservoir easy to handle without product deformation which results in less spillage
  • Reservoirs can be conveniently stacked with a unique nesting design that enables easy separation
  • Designed to stay flat on the benchtop and not warp
  • Sterile, sealed in bags and electron beam sterilised
  • DNase, RNase and pyrogen free
Graduation markings on the top edge and inner walls allows measurements of liquid to be easily identified and reduces waste of expensive reagents. Pour-off spouts design on all four corners enables easy spill-free pouring and reduces chance of dripping.
Certifications: Quality certificate available on request.
Packaging: Sealed in easy-to-open bags. No scissors needed to open.
Documents
Specifications
  • Sterile:
    -
  • Capacity:
    55 ml
  • Description:
    Reagent reservoir, PVC
  • Seq:
    03