Order Entry
Italy
ContactUsLinkComponent
Centrifuge tubes, 50 ml, GL 32, round bottom, nominal volume 45 ml, ungraduated, heavy wall, Ø 34x98 mm, with heat resistant screw c ap and PTFE - liner, borosilicate glass 3.3 1 * 1 items
Catalog # WITG4.945.203
Supplier:  witeg Labortechnik
Centrifuge tubes, 50 ml, GL 32, round bottom, nominal volume 45 ml, ungraduated, heavy wall, Ø 34x98 mm, with heat resistant screw c ap and PTFE - liner, borosilicate glass 3.3 1 * 1 items
Catalog # WITG4.945.203
Supplier:  witeg Labortechnik
Frequently Bought Together