Colour Standard Gardner 2 1 * 100 mL
Catalog # REGEGARD021
Supplier: REAGECON
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR®, pH-Indicator Paper Congo Red Reag. Ph. Eur. 1022002
Supplier: VWR Chemicals
N-Nitroso-ethyl-isopropylamine 100 µg/mL in Methanol 1 * 1 mL
Supplier: EHRENSTORFER
pH buffers, for LAQUAtwin meters
Supplier: HORIBA
Organic reference standard, Geosmin, Dr. Ehrenstorfer
Supplier: EHRENSTORFER
Flamprop-M-methyl certified 1 * 10 mg
Supplier: EHRENSTORFER
Iothalamic Acid, Reference Standard
Supplier: USP
Iridium standard solution, 1000 mg/L Ir in 20% hydrochloric acid (from IrCl₃), standard for ICP, Specpure®
Supplier: Thermo Fisher Scientific
Colour Standard Gardner 3 1 * 100 mL
Supplier: REAGECON
Ibuprofen, Supelco®
Supplier: Merck
Supelco® Organic Reference Standard, Dimethyl Terephthalate
Supplier: Merck
pH buffer solutions, referenced at 20 °C, traceable to SRM from NIST, Fluka™
Supplier: Honeywell Chemicals
Propylene oxide, Supelco®
Supplier: Merck
Supelco® Organic Reference Standard, Luteolin 7-glucoside
Supplier: Merck