200 FNU Turbidity Calibration solution for HI9829 1 * 1 items
Catalog # HANNHI9829-18
Supplier: Hanna
200 FNU Turbidity Calibration solution for HI9829 1 * 1 items
Catalog # HANNHI9829-18
Supplier: Hanna
Frequently Bought Together
pH buffers, ready to use
Supplier: WTW
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
OXYGEN SOLUTION ZERO DISSOLVED 1 * 1 L
Supplier: REAGECON