Order Entry
Italy
ContactUsLinkComponent
Screw In Fittings GL, For connection to BOLA Reactor Lids as GL necks. With hexagonal gripping surface in standard wrench size. 1 * 1 items
Codice catalogo BOHLB152-32
Supplier:  Bohlender
undefined
Screw In Fittings GL, For connection to BOLA Reactor Lids as GL necks. With hexagonal gripping surface in standard wrench size. 1 * 1 items
Codice catalogo BOHLB152-32
Supplier:  Bohlender
Frequently Bought Together