Columns, HiTrap™ Capto PlasmidSelect
HiTrap™ Capto PlasmidSelect, 7×25 mm, 1 ml, <4 ml/min
Catalog # 17-5499-11
Supplier: Cytiva
Jump to:
Product Details & Documents
Columns packed with Capto PlasmidSelect are suitable for purification of supercoiled DNA to high quality for gene therapy and DNA vaccine applications. Capto PlasmidSelect is a chromatography resin that allows supercoiled covalently closed circular forms of plasmid DNA to be separated from open circular forms.
- High-flow purification of supercoiled DNA from research scale to cGMP production
- Flexible process design due to a large operational window of flow velocities and bed heights
- Higher throughput for improved productivity and process economy
- Hydrophilic properties of base matrix prevent non-specific binding
- Bioprocess resin supported for industrial applications
- Convenient small-scale purification, process development, and scale-up using prepacked HiTrap columns
Capto PlasmidSelect is based on a rigid base matrix, delivering excellent pressure-flow properties to plasmid production.
Documents
Cytiva CyDye Licensing Statements Cytiva General Licensing Statements
Additional Product Details
Specifications
- Flow rate:<4 ml/min
- Capacity:1 ml
- Description:HiTrap™ Capto PlasmidSelect
- Bed dimensions:7×25 mm
- Cat. No.:17-5499-11
- Max. pressure:5 bar
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



