Thermometer, solid stem, similar to ASTM 21C, white backed, 79+87:0,1C, capillary tube specially coated inside, with non-wetting blue special liquid, total immersion, max.280x6-7mm, durable pigment, with Works Certificate with 2 test points 1 * 1 items
Codice catalogo AMARA300380-CB/WC
Supplier: Amarell
Frequently Bought Together