Order Entry
Italy
ContactUsLinkComponent
Thermometer similar to IP 32 C, solid stem, white backed, 98,6+101,4:0 ,05C, capillary tube specially coated inside, with non-wetting blue special liquid, total immersion, max.310x6-8mm, durable pigment, with ice point scale, calibrated with DKD Cer 1 * 1 items
Catalog # AMARA340430CBDKD3
Supplier:  Amarell
Thermometer similar to IP 32 C, solid stem, white backed, 98,6+101,4:0 ,05C, capillary tube specially coated inside, with non-wetting blue special liquid, total immersion, max.310x6-8mm, durable pigment, with ice point scale, calibrated with DKD Cer 1 * 1 items
Catalog # AMARA340430CBDKD3
Supplier:  Amarell
Frequently Bought Together