Order Entry
Italy
ContactUsLinkComponent
Fan, reinforced - 400 V option model, for increased air change rates, incl. uprated heat output for faster heating up time and improved recovery time max. 300 °C not in conjunction with load management option 1 * 1 items
Catalog # BIND8012-1062
Supplier:  Binder
Fan, reinforced - 400 V option model, for increased air change rates, incl. uprated heat output for faster heating up time and improved recovery time max. 300 °C not in conjunction with load management option 1 * 1 items
Catalog # BIND8012-1062
Supplier:  Binder
Frequently Bought Together