Order Entry
Italy
ContactUsLinkComponent
Conversion filter halogen, to convert the neutral white LED light to the more yellowish halogen type of light. To mount on the EasyLED Spotlight 1 * 1 items
Catalog # SCOC600690
Nome del fornitore:  SCHOTT AG Lighting and imaging
Conversion filter halogen, to convert the neutral white LED light to the more yellowish halogen type of light. To mount on the EasyLED Spotlight 1 * 1 items
Catalog # SCOC600690
Nome del fornitore:  SCHOTT AG Lighting and imaging
Frequently Bought Together