Solids Mix 2B solution for ICP-OES 6 components Al 5000mg/l Ca 5000mg/l Fe 5000mg/l K 2000mg/l Mg 5000mg/l Na 5000mg/l in HNO3 5%. Duree de conservation : 12 mois a ouverture du flacon, Duree de validite: 24 mois. 1 * 250 mL
Catalog # CMPLFC4C.5K.5N.L25
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together