6 components Mo 500mg/l Sb 1000mg/l Se 1000mg/l Sn 1000mg/l Te 1000mg/l Tl 1000mg/l in HNO3 5% HF 0.5%. Duree de conservation : 12 mois a ouverture du flacon, Duree de validite: 24 mois. 1 * 100 mL
Catalog # CMPL42F9.K55N05FL1
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together